Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | immunoglobulin kappa variable 4-59 | Igkv4-59 |
EG667683, Gm8760, kk4 |
-61.5kb | Anti-sense | Chr. 6 - 69438218 - 69438741 on NCBI Build GRCm38 |
Downstream | immunoglobulin kappa variable 4-57-1 | Igkv4-57-1 | Gm1418 | 43.8kb | Anti-sense | Chr. 6 - 69544368 - 69544892 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |