Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P124822 |
AK004017.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P124822 |
BC022628.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P124822 |
NM_001164210.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P124822 |
NM_133675.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
194604 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
676967 |
NM_146942.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
108744 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
612354 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
820707 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
265045 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
95107 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
922807 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
106392 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572720 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
853752 |
AK039909.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
734383 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
608097 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
588861 |
NM_008130.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
588861 |
X95255.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
458387 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
237779 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
527744 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
395621 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
885184 |
AK089969.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
554378 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
49755 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
2008 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
149339 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
67220 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P171660 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P120612 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P124822 |
AK004017.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P124822 |
BC022628.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P124822 |
NM_001164210.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P124822 |
NM_133675.2 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223238 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223266 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223247 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223225 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223242 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223252 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223260 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223239 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223265 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223236 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223234 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223237 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223253 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223251 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223254 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223269 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223249 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223255 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223231 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223257 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223230 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223264 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223268 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223244 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223261 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223228 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22223262 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P120612 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04942700 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P171660 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04942748 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04942713 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04942725 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P649276 |
AF365876.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P649276 |
AK004017.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P649276 |
AK010117.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P649276 |
BC022628.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P649276 |
NM_001164210.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P124822 |
AK004017.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P124822 |
BC022628.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P124822 |
NM_001164210.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P124822 |
NM_133675.2 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P120612 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P171660 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P04942725 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P04942700 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02392867 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02392874 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02352524 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02352516 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02392865 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223069 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223099 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223147 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223089 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223262 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223095 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223108 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223198 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223261 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223150 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223196 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223230 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223223 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223056 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223182 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223257 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223279 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223178 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223218 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223148 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223270 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223037 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223216 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223032 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223024 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223244 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223272 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223168 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223081 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223051 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223082 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223220 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223180 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223251 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223281 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223173 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223073 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223254 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223145 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223087 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223154 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223134 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223237 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223123 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223159 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223102 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223045 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223144 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223271 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223172 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223231 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223066 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223120 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223199 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223128 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223109 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223203 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223201 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223034 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223125 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223157 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223093 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223238 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223176 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223141 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223175 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223275 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223110 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223268 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223177 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223080 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223136 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223156 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223227 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223183 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223117 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223252 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223113 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223041 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223155 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223063 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223239 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223054 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223266 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223048 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223061 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223084 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223194 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223135 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223249 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223264 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223234 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223049 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223153 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223188 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223189 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223278 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223071 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223269 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223179 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223206 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223036 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223030 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223142 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223127 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223247 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223020 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223236 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223028 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223050 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223161 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223202 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223209 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223068 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223242 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223074 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223139 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223152 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22223064 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223276 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02392876 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02392872 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223159 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223225 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02954128 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223231 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02512401 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223216 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03062273 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223223 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223256 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223281 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223163 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223238 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223054 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223180 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02853436 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223089 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223095 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223175 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223257 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223279 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223247 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223227 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223244 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223048 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223057 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223059 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223152 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223271 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02579389 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223177 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223237 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223242 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02699528 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223278 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223261 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223178 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223269 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223172 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223251 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223051 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223264 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02352524 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223239 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223050 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223097 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223249 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223171 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02934851 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223093 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223260 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223148 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223155 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223262 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03062264 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02846962 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02856190 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02935449 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223228 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223252 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223056 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223179 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223150 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223170 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223268 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02699532 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02352522 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02999070 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223220 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223091 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223161 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223087 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223224 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223156 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02392873 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223092 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223218 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22223275 |
- |
- |