Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011952.2 | NP_036082.1 | 2 | 303 | 98 | |
GenBank mRNA | AK155287.1 | - | 1 | 245 | 98 | |
BC013754.1 | - | 2 | 136 | 98 | ||
BC029712.1 | - | 2 | 297 | 5041517 | 98 | |
BQ929318.1 | - | 2 | 303 | 98 | ||
Z14249.1 | - | 2 | 274 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon |