Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubtfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033793.3 | NP_001028965.1 | 3 | 1548 | 149 | |
GenBank mRNA | AK139937.1 | - | 3 | 1549 | 149 | |
AK163257.1 | - | 2 | 1665 | 149 | ||
BC139141.1 | - | 2 | 1363 | 8862163 | 149 | |
BC145822.1 | - | 2 | 1363 | 40131125 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |