Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039056.2 | NP_001034145.1 | 4 | 91 | ||
NM_001039057.2 | NP_001034146.1 | 4 | 91 | |||
NM_001271687.1 | NP_001258616.1 | 5 | 91 | |||
NM_001271689.1 | NP_001258618.1 | 5 | 91 | |||
NM_001271690.1 | NP_001258619.1 | 5 | 91 | |||
NM_001271691.1 | NP_001258620.1 | 4 | 91 | |||
NM_001271692.1 | NP_001258621.1 | 4 | 91 | |||
NM_001271693.1 | NP_001258622.1 | 4 | 91 | |||
NM_001271694.1 | NP_001258623.1 | 4 | 91 | |||
NM_001271695.1 | NP_001258624.1 | 3 | 91 | |||
NM_019664.5 | NP_062638.1 | 5 | 91 | |||
NR_073406.1 | 6 | 91 | ||||
GenBank mRNA | AK143004.1 | - | 4 | 91 | ||
AK165436.1 | - | 4 | 91 | |||
BC064098.1 | - | 2 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon |