Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ubiquitin-conjugating enzyme E2K | Ube2k |
AW492011, D5Ertd601e, E2-25k, Hip2, Hypg, Lig |
Sense | Chr. 5 - 65537238 - 65598989 on NCBI Build GRCm38 | |
Downstream |
PDS5, regulator of cohesion maintenance, homolog A (S. cerevisiae) |
Pds5a |
9030416H16Rik, E230024D05Rik, mKIAA0648 |
19.7kb | Anti-sense | Chr. 5 - 65608124 - 65698163 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |