Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap3s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF084575.1 | - | 1 - 2 | 148 | 105 | |
AK002565.1 | - | 1 - 2 | 174 | 105 | ||
AK137489.1 | - | 1 - 2 | 169 | 105 | ||
AK152100.1 | - | 1 - 2 | 180 | 105 | ||
BC012656.1 | - | 1 - 2 | 159 | 4187607 | 105 | |
BC108327.1 | - | 1 - 2 | 167 | 30713247 | 105 | |
BC131985.1 | - | 1 - 2 | 149 | 40130262 | 105 | |
BC145234.1 | - | 1 - 2 | 149 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |