Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap3s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF084575.1 | - | 4 - 5 | 419 | 158 | |
AK002565.1 | - | 4 - 5 | 445 | 158 | ||
AK137489.1 | - | 4 - 5 | 440 | 158 | ||
AK152100.1 | - | 4 - 5 | 451 | 158 | ||
AK194047.1 | - | 2 - 3 | 201 | 158 | ||
BC012656.1 | - | 4 - 5 | 430 | 4187607 | 158 | |
BC108327.1 | - | 4 - 5 | 438 | 30713247 | 158 | |
BC131985.1 | - | 4 - 5 | 420 | 40130262 | 158 | |
BC145234.1 | - | 4 - 5 | 420 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |