Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crybb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK020891.1 | - | 3 - 4 | 232 | 152 | |
BC085185.1 | - | 4 - 5 | 374 | 30679384 | 152 | |
M60559.1 | - | 2 - 3 | 199 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |