Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk1b22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010114.1 | NP_034244.1 | 2 - 3 | 219 | 122 | |
GenBank mRNA | BC106155.1 | - | 2 - 3 | 223 | 4189167 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |