Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcer1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010185.4 | NP_034315.1 | 1 - 2 | 156 | 121 | |
GenBank mRNA | AK155600.1 | - | 1 - 2 | 113 | 121 | |
BC034163.1 | - | 96 | 5065647 | 121 | ||
BY173505.1 | - | 1 - 2 | 156 | 121 | ||
CK791068.1 | - | 1 - 2 | 106 | 121 | ||
J05020.1 | - | 1 - 2 | 94 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |