Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900022206.mm.1 |
NM_205844.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022206.mm.1 |
NM_205844.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022206.mm.1 |
TC0900002685.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900022204.mm.1 |
NM_205844.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022204.mm.1 |
NM_205844.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022204.mm.1 |
TC0900002685.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900022205.mm.1 |
NM_205844.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022205.mm.1 |
NM_205844.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022205.mm.1 |
TC0900002685.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900022207.mm.1 |
NM_205844.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022207.mm.1 |
NM_205844.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022207.mm.1 |
TC0900002685.mm.1 |
MTA TC |