Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212485.2 | NP_997650.1 | 1 - 2 | 480 | 178 | |
GenBank mRNA | BC067067.1 | - | 1 - 2 | 480 | 30538572 | 178 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |