Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2141043 |
BQ175462.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2141043 |
AB164466.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2141043 |
NM_001011684.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
284557 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
470418 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
738078 |
BC100700.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
738078 |
BC100701.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
738078 |
BC100702.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
738078 |
BC100703.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
738078 |
NM_008570.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
735848 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
380150 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
537423 |
AK010793.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
537423 |
AK077427.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
279978 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
32975 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
163976 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P146537 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2141043 |
BQ175462.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2141043 |
AB164466.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2141043 |
NM_001011684.2 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162935 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162969 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162972 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162975 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162940 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162971 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162949 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162932 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162966 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162977 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162970 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162968 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162955 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162962 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162978 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162931 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162979 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162945 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162933 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162951 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162958 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162929 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162942 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162948 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162930 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162937 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162964 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162953 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162952 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20162974 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P146537 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04069952 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00049194 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P146537 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00366190 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00366198 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00366195 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00834432 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00366199 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163023 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163056 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162952 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163062 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163012 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162914 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163069 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163043 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163009 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163010 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162984 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162980 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163076 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162929 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162932 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162962 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163015 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162921 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163060 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162996 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163065 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163026 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162907 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162925 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163082 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163033 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163044 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162972 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162911 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162981 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162979 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162906 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162920 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163073 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163036 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162966 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162995 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163039 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163080 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163059 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163031 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162970 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162942 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162931 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162935 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163017 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162918 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163067 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162930 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162948 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162994 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162971 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163046 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162945 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163021 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162975 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163005 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162998 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163041 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162919 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162908 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162990 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163008 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163063 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163040 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162974 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162968 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162940 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163049 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162964 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163078 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163001 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162957 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163075 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163018 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162912 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162969 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163028 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163047 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20162977 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163019 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20163071 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162950 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20163014 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162926 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162966 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162970 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01000150 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162972 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00748165 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01000144 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162957 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162981 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162977 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162945 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20163066 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162964 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01137573 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162948 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162931 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00595160 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01053458 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162930 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00595166 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20163008 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20163005 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00385519 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20163062 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162980 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00664883 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00385515 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00944523 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00944531 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20163063 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20163078 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00834432 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00664888 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162953 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00476780 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162987 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00153222 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00256712 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00595163 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162974 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00748166 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01000147 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162940 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162969 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162984 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162932 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162933 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00944525 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162995 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162962 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162975 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01000154 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162978 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20163075 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20163000 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01057564 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00595162 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162937 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00366201 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00944519 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162952 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00069501 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162942 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162996 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00850135 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20162968 |
- |
- |