Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mthfs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026829.2 | NP_081105.1 | 2 | 364 | 131 | |
GenBank mRNA | AK004092.1 | - | 2 | 365 | 131 | |
AK154942.1 | - | 3 | 1037 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Mus musculus Amplicon spans exons and probe does not span exons |