Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P405304 |
NM_053172.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P405304 |
AF298661.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P405304 |
AF314964.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P405304 |
AK158231.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P405304 |
NM_053172.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P405304 |
AF298661.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P405304 |
AF314964.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P405304 |
AK158231.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
288840 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
634105 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454301 |
AK006421.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454301 |
AK007536.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454301 |
AK009312.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454301 |
AK028010.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454301 |
AK028397.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454301 |
AK145695.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454301 |
BC064674.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454301 |
NM_025454.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
432053 |
AK016710.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
35591 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
55279 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
194665 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
70751 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P110135 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P139188 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P405304 |
NM_053172.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P405304 |
AF298661.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P405304 |
AF314964.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P405304 |
AK158231.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P405304 |
NM_053172.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P405304 |
AF298661.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P405304 |
AF314964.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P405304 |
AK158231.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P405304 |
NM_053172.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P405304 |
AF298661.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P405304 |
AF314964.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P405304 |
AK158231.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P405304 |
NM_053172.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P405304 |
AF298661.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P405304 |
AF314964.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P405304 |
AK158231.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951592 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951563 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951569 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951596 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951601 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951594 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951566 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951565 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951572 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951570 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951589 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951580 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951568 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951577 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951585 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951587 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951608 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951586 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951607 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951582 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951605 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951579 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951564 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951595 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951609 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951600 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951573 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951571 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951591 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951598 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23951604 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01177953 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P139188 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P405304 |
NM_053172.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P405304 |
AF298661.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P405304 |
AF314964.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P405304 |
AK158231.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P405304 |
NM_053172.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P405304 |
AF298661.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P405304 |
AF314964.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P405304 |
AK158231.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P104188 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P139188 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04104840 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04374583 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04104839 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04104838 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04104853 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951672 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951564 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951634 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951667 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951611 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951632 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951608 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951677 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951637 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951621 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951622 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951719 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951625 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951711 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951684 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951715 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951642 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951651 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951675 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951709 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951628 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951671 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951616 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951723 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951636 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951587 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951566 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951727 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951730 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951585 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951662 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951687 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951721 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951713 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951700 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951598 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951664 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951600 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951717 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951648 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951579 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951624 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951644 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951722 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951605 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951697 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951660 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951650 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951668 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951563 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951646 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951571 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951591 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951577 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951689 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951704 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951725 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951676 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951570 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951716 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951618 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951589 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951640 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951629 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951682 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951573 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951657 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951688 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951613 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951690 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951568 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951686 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951594 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951706 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951592 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951572 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951679 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951565 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951638 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951586 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951615 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951685 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951702 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23951580 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951625 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951566 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951621 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04514473 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04984052 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951589 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951613 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951564 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951592 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951616 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951587 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951618 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951596 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951565 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04530527 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951591 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951632 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951570 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951637 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951573 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951581 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04984066 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951615 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951569 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951611 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951568 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951638 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04374583 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04319385 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951594 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04530524 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951620 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951639 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951605 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04948090 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951585 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951600 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951629 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04984058 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04319360 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951586 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04530534 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951561 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951563 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951579 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04319363 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951580 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951624 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951598 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04319375 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23951577 |
- |
- |