Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK020445.1 | - | 2 | 1033 | 168 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ubiquitin-associated protein 1 | Ubap1 |
2700092A01Rik, NAG20, UBAP-1, Ubap |
-64.6kb | Anti-sense | Chr. 4 - 41348996 - 41389766 on NCBI Build GRCm38 |
Downstream | nudix (nucleoside diphosphate linked moiety X)-type motif 2 | Nudt2 |
2310051L06Rik, AA939917, APAH1 |
9.7kb | Anti-sense | Chr. 4 - 41465080 - 41480926 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |