Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phkg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011079.2 | NP_035209.1 | 7 - 8 | 820 | 75 | |
GenBank mRNA | BC055102.1 | - | 7 - 8 | 820 | 6532722 | 75 |
J03293.1 | - | 7 - 8 | 690 | 75 | ||
M16216.1 | - | 6 - 7 | 658 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |