Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017966.2 | NP_001017966.1 | 4 - 5 | 855 | 75 | |
GenBank mRNA | AK129015.1 | - | 833 | 75 | ||
BC021415.1 | - | 4 - 5 | 836 | 75 | ||
BF468816.1 | - | 3 - 4 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |