Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017966.2 | NP_001017966.1 | 5 - 6 | 984 | 62 | |
GenBank mRNA | AK129015.1 | - | 962 | 62 | ||
BC021415.1 | - | 5 - 6 | 965 | 62 | ||
BF468816.1 | - | 2 - 3 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |