Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033323.3 | NP_001028495.2 | 12 - 13 | 1861 | 81 | |
NM_001129787.1 | NP_001123259.1 | 12 - 13 | 1861 | 81 | ||
GenBank mRNA | AK158950.1 | - | 12 - 13 | 1861 | 81 | |
BC150882.1 | - | 12 - 13 | 1669 | 81 | ||
BC150887.1 | - | 12 - 13 | 1669 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |