Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033397.5 | NP_001028569.2 | 1 - 2 | 493 | 67 | |
GenBank mRNA | AK028591.1 | - | 1 - 2 | 493 | 67 | |
AK028682.1 | - | 1 - 2 | 493 | 67 | ||
AK132445.1 | - | 1 - 2 | 497 | 67 | ||
AK162718.1 | - | 1 - 2 | 495 | 67 | ||
BC116672.1 | - | 1 - 2 | 482 | 40107596 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |