Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nutm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033400.2 | NP_001028572.1 | 3 - 4 | 975 | 79 | |
GenBank mRNA | AK133253.1 | - | 3 - 4 | 976 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Mus musculus Probe spans exons |