Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033408.4 | NP_001028580.1 | 11 - 12 | 2495 | 124 | |
GenBank mRNA | AK133355.1 | - | 11 - 12 | 2496 | 124 | |
BC150950.1 | - | 11 - 12 | 2457 | 9087861 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |