Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033775.3 | NP_001028947.1 | 4 - 5 | 866 | 82 | |
GenBank mRNA | AK133415.1 | - | 4 - 5 | 867 | 82 | |
BC147608.1 | - | 4 - 5 | 884 | 9056598 | 82 | |
BC157912.1 | - | 4 - 5 | 884 | 9088129 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |