Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039388.2 | NP_001034477.1 | 2 - 3 | 423 | 87 | |
NM_001039389.2 | NP_001034478.1 | 2 - 3 | 344 | 87 | ||
NM_172445.3 | NP_766033.1 | 2 - 3 | 344 | 87 | ||
GenBank mRNA | AK014128.1 | - | 2 - 3 | 320 | 87 | |
AK032582.1 | - | 2 - 3 | 306 | 87 | ||
AK036203.1 | - | 2 - 3 | 340 | 87 | ||
AK044740.1 | - | 2 - 3 | 324 | 87 | ||
AK129254.1 | - | 2 - 3 | 300 | 87 | ||
AK146209.1 | - | 2 - 3 | 319 | 87 | ||
BB611667.1 | - | 2 - 3 | 423 | 87 | ||
BC046236.1 | - | 1 - 2 | 171 | 6817089 | 87 | |
BC048819.1 | - | 2 - 3 | 268 | 87 | ||
CJ134699.1 | - | 2 - 3 | 345 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |