Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039388.2 | NP_001034477.1 | 4 - 5 | 630 | 82 | |
NM_001039389.2 | NP_001034478.1 | 4 - 5 | 551 | 82 | ||
NM_172445.3 | NP_766033.1 | 4 - 5 | 551 | 82 | ||
GenBank mRNA | AK014128.1 | - | 4 - 5 | 527 | 82 | |
AK032582.1 | - | 4 - 5 | 513 | 82 | ||
AK036203.1 | - | 4 - 5 | 547 | 82 | ||
AK044740.1 | - | 4 - 5 | 531 | 82 | ||
AK129254.1 | - | 4 - 5 | 507 | 82 | ||
AK146209.1 | - | 4 - 5 | 526 | 82 | ||
BC046236.1 | - | 3 - 4 | 378 | 6817089 | 82 | |
BC048819.1 | - | 4 - 5 | 475 | 82 | ||
CJ134699.1 | - | 4 - 5 | 552 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |