Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011296.2 | NP_035426.1 | 2 - 3 | 127 | 74 | |
GenBank mRNA | AK050626.1 | - | 2 - 3 | 150 | 74 | |
AK144398.1 | - | 74 | ||||
AK146354.1 | - | 2 - 3 | 150 | 74 | ||
AK151580.1 | - | 2 - 3 | 150 | 74 | ||
AK169246.1 | - | 2 - 3 | 150 | 74 | ||
BC081458.1 | - | 1 - 2 | 115 | 5712669 | 74 | |
BC081459.1 | - | 2 - 3 | 127 | 6407328 | 74 | |
BC093500.1 | - | 2 - 3 | 128 | 4207659 | 74 | |
BC100604.1 | - | 2 - 3 | 149 | 30120583 | 74 | |
M76763.1 | - | 2 - 3 | 128 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |