Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppih | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110129.1 | NP_001103599.1 | 4 - 5 | 249 | 117 | |
NM_001110130.1 | NP_001103600.1 | 4 - 5 | 249 | 117 | ||
NM_028677.4 | NP_082953.1 | 4 - 5 | 249 | 117 | ||
GenBank mRNA | AK003179.1 | - | 4 - 5 | 227 | 117 | |
AK005202.1 | - | 4 - 5 | 210 | 117 | ||
AK008394.1 | - | 4 - 5 | 227 | 117 | ||
AK014665.1 | - | 4 - 5 | 210 | 117 | ||
AK134995.1 | - | 4 - 5 | 229 | 117 | ||
AK136330.1 | - | 4 - 5 | 208 | 117 | ||
AK164299.1 | - | 4 - 5 | 214 | 117 | ||
BC016565.1 | - | 4 - 5 | 204 | 117 | ||
BC050116.1 | - | 4 - 5 | 203 | 6432881 | 117 | |
BC098490.1 | - | 205 | 30022355 | 117 | ||
BY708305.1 | - | 4 - 5 | 226 | 117 | ||
CJ071503.1 | - | 4 - 5 | 272 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |