Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrgpra5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207534.1 | NP_997417.1 | 607 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Recommended Assay Mus musculus Inventoried |