Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700005394.mm.1 |
NM_001002842.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005394.mm.1 |
NM_001002842.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005394.mm.1 |
NR_028279.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005394.mm.1 |
NR_028279.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005394.mm.1 |
TC1700002776.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700005398.mm.1 |
NM_001002842.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005398.mm.1 |
NM_001002842.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005398.mm.1 |
NR_028279.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005398.mm.1 |
NR_028279.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005398.mm.1 |
TC1700002776.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700005393.mm.1 |
NM_001002842.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005393.mm.1 |
NM_001002842.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005393.mm.1 |
TC1700002776.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700005396.mm.1 |
NR_028279.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005396.mm.1 |
NR_028279.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005396.mm.1 |
TC1700002776.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700005395.mm.1 |
NM_001002842.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005395.mm.1 |
NM_001002842.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005395.mm.1 |
NR_028279.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005395.mm.1 |
NR_028279.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005395.mm.1 |
TC1700002776.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700005399.mm.1 |
NM_001002842.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005399.mm.1 |
NM_001002842.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005399.mm.1 |
NR_028279.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005399.mm.1 |
NR_028279.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005399.mm.1 |
TC1700002776.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700005397.mm.1 |
NM_001002842.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005397.mm.1 |
NM_001002842.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005397.mm.1 |
NR_028279.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005397.mm.1 |
NR_028279.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005397.mm.1 |
TC1700002776.mm.1 |
MTA TC |