Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027261.3 | NP_081537.1 | 4 - 5 | 722 | 85 | |
NM_029248.2 | NP_083524.2 | 4 - 5 | 722 | 85 | ||
NR_028401.1 | 4 - 5 | 722 | 85 | |||
GenBank mRNA | AK012656.1 | - | 2 - 3 | 572 | 85 | |
AK012662.1 | - | 4 - 5 | 674 | 85 | ||
AK014920.1 | - | 4 - 5 | 674 | 85 | ||
AK016109.1 | - | 4 - 5 | 676 | 85 | ||
AK050165.1 | - | 4 - 5 | 678 | 85 | ||
AK161656.1 | - | 4 - 5 | 677 | 85 | ||
AK166329.1 | - | 4 - 5 | 677 | 85 | ||
AK166633.1 | - | 4 - 5 | 677 | 85 | ||
BC027813.1 | - | 2 - 3 | 510 | 85 | ||
BC056964.1 | - | 3 - 4 | 660 | 85 | ||
BC110660.1 | - | 3 - 4 | 664 | 3158156 | 85 | |
BC117068.1 | - | 3 - 4 | 654 | 40126387 | 85 | |
BC117070.1 | - | 3 - 4 | 654 | 40126389 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |