Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700033009.mm.1 |
NR_033673.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033009.mm.1 |
NR_033673.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033009.mm.1 |
TC0700003950.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700033010.mm.1 |
NR_033673.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033010.mm.1 |
NR_033673.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033010.mm.1 |
TC0700003950.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700033007.mm.1 |
NR_033673.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033007.mm.1 |
NR_033673.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033007.mm.1 |
TC0700003950.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700033008.mm.1 |
NR_033673.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033008.mm.1 |
NR_033673.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033008.mm.1 |
TC0700003950.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700033011.mm.1 |
NR_033673.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033011.mm.1 |
NR_033673.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033011.mm.1 |
TC0700003950.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700033005.mm.1 |
NR_033673.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033005.mm.1 |
NR_033673.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033005.mm.1 |
TC0700003950.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700033006.mm.1 |
NR_033673.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033006.mm.1 |
NR_033673.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033006.mm.1 |
TC0700003950.mm.1 |
MTA TC |