Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P139052 |
AK143822.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P139052 |
BC147284.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P139052 |
BC147285.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P139052 |
NM_001039038.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2028288 |
AK143822.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2028288 |
NM_001039038.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855531 |
AF099067.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855531 |
AJ223071.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855531 |
AK078809.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855531 |
AK132803.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855531 |
AK147381.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855531 |
AK149147.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855531 |
AK149584.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855531 |
BC057939.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855531 |
NM_011849.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855531 |
Y09234.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
155378 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
14619 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P139052 |
AK143822.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P139052 |
BC147284.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P139052 |
BC147285.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P139052 |
NM_001039038.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2028288 |
AK143822.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2028288 |
NM_001039038.2 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107387 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107389 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107377 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107402 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107411 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107353 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107348 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107407 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107374 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107403 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107365 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107410 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107352 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107358 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107363 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107356 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107371 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107413 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107385 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107366 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107418 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107361 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107364 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107350 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107367 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107404 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107399 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107392 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107368 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107416 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107347 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107409 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107400 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107391 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107397 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107412 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107395 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107376 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107401 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107393 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107349 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107380 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107378 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107386 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107382 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107394 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31107369 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13483411 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13483405 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13483406 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13483425 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13135566 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13422586 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107296 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107275 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107394 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107407 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107327 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107397 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107371 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107372 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107331 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107309 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107303 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107382 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107339 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107301 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107392 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107431 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107347 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107288 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107422 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107299 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107328 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107374 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107418 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107356 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107412 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107389 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107364 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107365 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107380 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107332 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107363 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107434 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107284 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107386 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107409 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107439 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107425 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107291 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107341 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107378 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107352 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107405 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107289 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107286 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107311 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107330 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107417 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107410 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107305 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107413 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107354 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107428 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107436 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107293 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107295 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107316 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107430 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107335 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107366 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107361 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107424 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107369 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107368 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107411 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107421 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107367 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107353 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107420 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107358 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107391 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107376 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107314 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107402 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107323 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107350 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107348 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107437 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107318 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107403 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107432 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107377 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107307 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107338 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107312 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107399 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31107343 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13152974 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107437 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107403 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13483418 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107382 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107402 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13422582 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107395 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107410 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107392 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107420 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107377 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107407 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107372 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107400 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13152962 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13501873 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107393 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107347 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13471624 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107358 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107361 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13152951 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107436 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13152979 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107380 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13386795 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13558577 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107404 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13483425 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107441 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107438 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107365 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107378 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107350 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107425 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107397 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107356 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13483416 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107439 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107430 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107428 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107369 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13519006 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107413 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13152954 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107405 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107376 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107431 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107364 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13483424 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107353 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13518968 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107363 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107371 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107354 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13483413 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107412 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13152963 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31107417 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13558581 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13152984 |
- |
- |