Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10429572 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10429576 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10429570 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10429574 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10429569 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10429576 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10429569 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10429571 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10429574 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10429570 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10429577 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10429573 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10429568 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10429573 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10429568 |
- |
- |
Mouse Genome 430 2.0 Array |
1421571_PM_a_at |
- |
- |
Mouse Genome 430 2.0 Array |
1421571_a_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1500013048.mm.1 |
NM_001252055.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013048.mm.1 |
NM_001252055.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013048.mm.1 |
NM_010741.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013048.mm.1 |
NM_010741.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013048.mm.1 |
TC1500001729.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500013045.mm.1 |
NM_001252056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013045.mm.1 |
NM_001252056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013045.mm.1 |
TC1500001729.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500013052.mm.1 |
NM_001252055.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013052.mm.1 |
NM_001252055.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013052.mm.1 |
NM_001252056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013052.mm.1 |
NM_001252056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013052.mm.1 |
NM_001252057.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013052.mm.1 |
NM_001252057.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013052.mm.1 |
NM_001252058.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013052.mm.1 |
NM_001252058.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013052.mm.1 |
NM_010741.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013052.mm.1 |
NM_010741.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013052.mm.1 |
TC1500001729.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500013059.mm.1 |
NM_001099217.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013059.mm.1 |
NM_001099217.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013059.mm.1 |
TC1500001730.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500013049.mm.1 |
NM_001252055.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013049.mm.1 |
NM_001252055.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013049.mm.1 |
NM_001252056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013049.mm.1 |
NM_001252056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013049.mm.1 |
NM_001252057.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013049.mm.1 |
NM_001252057.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013049.mm.1 |
NM_001252058.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013049.mm.1 |
NM_001252058.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013049.mm.1 |
NM_010741.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013049.mm.1 |
NM_010741.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013049.mm.1 |
TC1500001729.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500013060.mm.1 |
NM_001099217.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013060.mm.1 |
NM_001099217.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013060.mm.1 |
TC1500001730.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500013044.mm.1 |
NM_001252056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013044.mm.1 |
NM_001252056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013044.mm.1 |
TC1500001729.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500013055.mm.1 |
NM_001252055.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013055.mm.1 |
NM_001252055.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013055.mm.1 |
NM_001252056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013055.mm.1 |
NM_001252056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013055.mm.1 |
NM_001252057.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013055.mm.1 |
NM_001252057.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013055.mm.1 |
NM_010741.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013055.mm.1 |
NM_010741.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013055.mm.1 |
TC1500001729.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500013051.mm.1 |
NM_001252055.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013051.mm.1 |
NM_001252055.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013051.mm.1 |
NM_001252056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013051.mm.1 |
NM_001252056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013051.mm.1 |
NM_001252057.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013051.mm.1 |
NM_001252057.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013051.mm.1 |
NM_001252058.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013051.mm.1 |
NM_001252058.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013051.mm.1 |
NM_010741.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013051.mm.1 |
NM_010741.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013051.mm.1 |
TC1500001729.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500013058.mm.1 |
NM_001099217.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013058.mm.1 |
NM_001099217.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013058.mm.1 |
TC1500001730.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1421571_PM_a_at |
- |
- |
Mouse Expression Set 430 |
1421571_PM_a_at |
- |
- |
Mouse Expression Set 430 |
1421571_a_at |
- |
- |
Mouse Expression Set 430 |
1421571_a_at |
- |
- |
Murine Genome U74v2 Set |
93077_s_at |
- |
- |
Murine Genome U74v2 Set |
93077_s_at |
- |
- |
Murine Genome U74v2 Set |
93077_PM_s_at |
- |
- |
Murine Genome U74v2 Set |
93077_PM_s_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1421571_PM_a_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1421571_a_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1421571_PM_a_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1421571_a_at |
- |
- |