Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026034.4 | NP_080310.1 | 4 - 5 | 718 | 74 | |
GenBank mRNA | AK009821.1 | - | 4 - 5 | 712 | 74 | |
AK011303.1 | - | 4 - 5 | 717 | 74 | ||
AK015272.1 | - | 4 - 5 | 851 | 74 | ||
BC027343.1 | - | 4 - 5 | 649 | 74 | ||
BC038487.1 | - | 4 - 5 | 670 | 2812558 | 74 | |
BC046281.1 | - | 4 - 5 | 670 | 6395384 | 74 | |
BC058573.1 | - | 4 - 5 | 759 | 6487440 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |