Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfwd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146218.4 | NP_666330.2 | 6 - 7 | 1125 | 96 | |
GenBank mRNA | AK137499.1 | - | 6 - 7 | 1105 | 96 | |
AK137698.1 | - | 6 - 7 | 1088 | 96 | ||
AK139032.1 | - | 6 - 7 | 1106 | 96 | ||
BC018533.1 | - | 6 - 7 | 1094 | 2651526 | 96 | |
BC023694.1 | - | 6 - 7 | 1130 | 5344528 | 96 | |
BC027246.1 | - | 1 - 2 | 76 | 3497792 | 96 | |
BC096602.1 | - | 6 - 7 | 1088 | 4954068 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |