Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Net1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047159.2 | NP_001040624.1 | 8 - 9 | 1233 | 119 | |
NM_019671.3 | NP_062645.2 | 10 - 11 | 1354 | 119 | ||
GenBank mRNA | AJ010045.1 | - | 8 - 9 | 1196 | 119 | |
AK082128.1 | - | 8 - 9 | 1230 | 119 | ||
AK088632.1 | - | 8 - 9 | 1197 | 119 | ||
AK139921.1 | - | 8 - 9 | 1197 | 119 | ||
BC004699.1 | - | 8 - 9 | 1207 | 3499258 | 119 | |
CA980605.1 | - | 4 - 5 | 536 | 119 | ||
CN697204.1 | - | 2 - 3 | 353 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |