Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146348.1 | NP_001139820.1 | 7 - 8 | 1351 | 130 | |
NM_001146350.1 | NP_001139822.1 | 7 - 8 | 1354 | 130 | ||
NM_007932.2 | NP_031958.2 | 7 - 8 | 1354 | 130 | ||
GenBank mRNA | AK151302.1 | - | 7 - 8 | 1324 | 130 | |
AY679531.1 | - | 7 - 8 | 1354 | 130 | ||
BC029080.1 | - | 7 - 8 | 1268 | 4168250 | 130 | |
S69407.1 | - | 8 - 9 | 1219 | 130 | ||
X77952.1 | - | 7 - 8 | 1354 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |