Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Macrod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028387.1 | NP_082663.1 | 4 | 652 | 57 | |
GenBank mRNA | AK004072.1 | - | 4 | 609 | 57 | |
AK029184.1 | - | 4 | 632 | 57 | ||
BC027552.1 | - | 4 | 446 | 57 | ||
BC028443.1 | - | 4 | 446 | 57 | ||
BC038317.1 | - | 4 | 578 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon |