Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdh9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081377.2 | NP_001074846.1 | 2 - 3 | 3281 | 105 | |
GenBank mRNA | AY861423.1 | - | 2 - 3 | 617 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Inventoried Probe spans exons |