Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081435.1 | NP_001074904.1 | 8 - 9 | 1196 | 86 | |
GenBank mRNA | BC158049.1 | - | 8 - 9 | 1131 | 9007353 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |