Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081633.1 | NP_001075102.1 | 3 - 4 | 450 | 81 | |
GenBank mRNA | AK088520.1 | - | 3 - 4 | 415 | 81 | |
BC013711.1 | - | 3 - 4 | 429 | 3708646 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |