Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080387.2 | NP_001073856.1 | 5 - 6 | 651 | 89 | |
NM_001284195.1 | NP_001271124.1 | 5 - 6 | 648 | 89 | ||
GenBank mRNA | AB015894.1 | - | 5 - 6 | 562 | 89 | |
AF060490.1 | - | 5 - 6 | 586 | 89 | ||
AK014345.1 | - | 5 - 6 | 590 | 89 | ||
AK134289.1 | - | 5 - 6 | 621 | 89 | ||
AK135316.1 | - | 5 - 6 | 596 | 89 | ||
AK168524.1 | - | 5 - 6 | 609 | 89 | ||
AK168558.1 | - | 5 - 6 | 609 | 89 | ||
BC006670.1 | - | 5 - 6 | 603 | 89 | ||
BC037591.1 | - | 5 - 6 | 569 | 5004611 | 89 | |
BC043060.1 | - | 5 - 6 | 975 | 6419883 | 89 | |
BC083082.1 | - | 5 - 6 | 563 | 6511807 | 89 | |
CF914989.1 | - | 5 - 6 | 564 | 89 | ||
CN528501.1 | - | 4 - 5 | 350 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |