Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080933.1 | NP_001074402.1 | 5 - 6 | 871 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |