Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Larp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024526.2 | NP_001019697.2 | 2 - 3 | 341 | 112 | |
NM_001080948.2 | NP_001074417.1 | 2 - 3 | 354 | 112 | ||
NM_001284521.1 | NP_001271450.1 | 2 - 3 | 328 | 112 | ||
NM_001284522.1 | NP_001271451.1 | 2 - 3 | 351 | 112 | ||
NM_001284523.1 | NP_001271452.1 | 2 - 3 | 344 | 112 | ||
GenBank mRNA | AK049756.1 | - | 2 - 3 | 268 | 112 | |
AK139636.1 | - | 2 - 3 | 347 | 112 | ||
AK139909.1 | - | 2 - 3 | 192 | 112 | ||
AK165392.1 | - | 2 - 3 | 289 | 112 | ||
AK172198.1 | - | 2 - 3 | 286 | 112 | ||
BC075726.1 | - | 2 - 3 | 256 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |