Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aurkc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080965.1 | NP_001074434.1 | 2 - 3 | 199 | 120 | |
GenBank mRNA | BC100337.1 | - | 2 - 3 | 120 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |