Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hba-x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010405.4 | NP_034535.1 | 1 - 2 | 162 | 93 | |
GenBank mRNA | AK077590.1 | - | 1 - 2 | 161 | 93 | |
BC051988.1 | - | 1 - 2 | 132 | 5711898 | 93 | |
BY111754.1 | - | 1 - 2 | 163 | 93 | ||
M26898.1 | - | 1 - 2 | 120 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons |