Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199059.1 | NP_001185988.1 | 6 - 7 | 800 | 159 | |
NM_019983.2 | NP_064367.1 | 5 - 6 | 945 | 159 | ||
GenBank mRNA | AF093590.1 | - | 6 - 7 | 776 | 159 | |
AK044568.1 | - | 5 - 6 | 965 | 159 | ||
AK146734.1 | - | 6 - 7 | 750 | 159 | ||
BC018229.1 | - | 5 - 6 | 945 | 4507477 | 159 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |