Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024919.1 | NP_001020090.1 | 1 - 2 | 358 | 121 | |
GenBank mRNA | AK009485.1 | - | 1 - 2 | 340 | 121 | |
BB616782.1 | - | 1 - 2 | 382 | 121 | ||
BC116435.1 | - | 2 - 3 | 190 | 40104481 | 121 | |
BC116436.1 | - | 2 - 3 | 190 | 40104482 | 121 | |
BC128508.1 | - | 2 - 3 | 260 | 121 | ||
BC128509.1 | - | 2 - 3 | 190 | 40104485 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |